(2Z)-[(2-Ethoxy-2-oxoethoxy)imino](1H-pyrazol-5-yl)acetic acid

Supplier: Matrix Scientific

080682-500MG 080682-1G 080682-5G
102670-750EA 258.46 USD
102670-750 102670-752 102670-754
(2Z)-[(2-Ethoxy-2-oxoethoxy)imino](1H-pyrazol-5-yl)acetic acid
(2Z)-[(2-Ethoxy-2-oxoethoxy)imino](1H-pyrazol-5-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₅
MDL Number: MFCD01026129
CAS Number: 76674-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR