2-Hydroxy-3,5-dinitropyridine

Supplier: Matrix Scientific

080681-500MG 080681-1G
102670-746EA 387.69 USD
102670-746 102670-748
2-Hydroxy-3,5-dinitropyridine
2-Hydroxy-3,5-dinitropyridine

Hazard: Irritant

Formula: C₅H₃N₃O₅
MDL Number: MFCD00039716
CAS Number: 2980-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR