Morpholine-3,5-dione

Supplier: Matrix Scientific

080679-500MG 080679-1G
102670-740EA 467.69 USD
102670-740 102670-742
Morpholine-3,5-dione
Morpholine-3,5-dione

Hazard: Irritant

Formula: C₄H₅NO₃
MDL Number: MFCD01863549
CAS Number: 4430-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR