3,5-Dimethyl-1H-pyrazol-4-ol

Supplier: Matrix Scientific

080675-500MG 080675-1G 080675-5G
102670-718EA 387.69 USD
102670-718 102670-720 102670-722
3,5-Dimethyl-1H-pyrazol-4-ol
3,5-Dimethyl-1H-pyrazol-4-ol

Hazard: Irritant

Formula: C₅H₈N₂O
MDL Number: MFCD00030421
CAS Number: 2820-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR