2-Chloro-8-methoxyquinoline

Supplier: Matrix Scientific

080673-1G 080673-5G
102670-708EA 361.54 USD
102670-708 102670-710
2-Chloro-8-methoxyquinoline
2-Chloro-8-methoxyquinoline

Hazard: Irritant

Formula: C₁₀H₈ClNO
MDL Number: MFCD02684171
CAS Number: 74668-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR