8-(Benzyloxy)quinoline-2-carbaldehyde

Supplier: Matrix Scientific

080669-500MG 080669-1G 080669-5G
102670-682EA 387.69 USD
102670-682 102670-684 102670-686
8-(Benzyloxy)quinoline-2-carbaldehyde
8-(Benzyloxy)quinoline-2-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₃NO₂
MDL Number: MFCD00636990
CAS Number: 88238-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR