7-Methylquinolin-8-ol

Supplier: Matrix Scientific

080666-500MG 080666-1G 080666-5G
102670-664EA 646.15 USD
102670-664 102670-666 102670-668
7-Methylquinolin-8-ol
7-Methylquinolin-8-ol

Hazard: Irritant

Formula: C₁₀H₉NO
MDL Number: MFCD00168979
CAS Number: 5541-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR