5,7-Dimethyl-8-quinolinol

Supplier: Matrix Scientific

Synonyms: 5,7-Dimethyl-8-hydroxyquinoline

080665-500MG 080665-1G 080665-5G
102670-658EA 387.69 USD
102670-658 102670-660 102670-662
5,7-Dimethyl-8-quinolinol
5,7-Dimethyl-8-quinolinol

Hazard: Irritant

Formula: C₁₁H₁₁NO
MW: 173.2111 g/mol
MDL Number: MFCD00168976
CAS Number: 37873-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR