8-Hydroxyquinoline-2-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 8-Hydroxyquinoline-2-carboxaldehyde

080662-500MG 080662-1G 080662-5G
102670-640EA 193.85 USD
102670-640 102670-642 102670-644
8-Hydroxyquinoline-2-carbaldehyde
8-Hydroxyquinoline-2-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
MDL Number: MFCD00168962
CAS Number: 14510-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR