2-Chloroquinolin-8-ol

Supplier: Matrix Scientific

080661-500MG 080661-1G 080661-5G
102670-634EA 258.46 USD
102670-634 102670-636 102670-638
2-Chloroquinolin-8-ol
2-Chloroquinolin-8-ol

Hazard: Irritant

Formula: C₉H₆ClNO
MDL Number: MFCD00168950
CAS Number: 31568-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR