2-Piperidin-1-ylquinolin-8-ol

Supplier: Matrix Scientific

080658-500MG 080658-1G 080658-5G
102670-616EA 258.46 USD
102670-616 102670-618 102670-620
2-Piperidin-1-ylquinolin-8-ol
2-Piperidin-1-ylquinolin-8-ol

Hazard: Irritant

Formula: C₁₄H₁₆N₂O
MDL Number: MFCD00168935
CAS Number: 31570-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR