2-(3-Bromo-1H-1,2,4-triazol-5-yl)pyridine

Supplier: Matrix Scientific

080654-500MG 080654-1G 080654-5G
102670-598EA 387.69 USD
102670-598 102670-600 102670-602
2-(3-Bromo-1H-1,2,4-triazol-5-yl)pyridine
2-(3-Bromo-1H-1,2,4-triazol-5-yl)pyridine

Hazard: Irritant

Formula: C₇H₅BrN₄
MDL Number: MFCD13188572
CAS Number: 219508-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR