3-Pyridyl acetate

Supplier: Matrix Scientific

Synonyms: Pyridin-3-yl acetate, 3-Acetoxypyridine, 3-Pyridyl acetate

080651-500MG 080651-1G 080651-5G
102670-582EA 161.54 USD
102670-582 102670-584 102670-586
3-Pyridyl acetate
3-Pyridyl acetate

Hazard: Irritant

Formula: C₇H₇NO₂
MDL Number: MFCD00006377
CAS Number: 17747-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR