3-Amino-1-(4-nitrophenyl)-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

080644-500MG 080644-1G 080644-5G
102670-546EA 387.69 USD
102670-546 102670-548 102670-550
3-Amino-1-(4-nitrophenyl)-1H-pyrazole-4-carbonitrile
3-Amino-1-(4-nitrophenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₀H₇N₅O₂
MDL Number: MFCD00828837
CAS Number: 219930-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR