3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

080643-500MG 080643-1G 080643-5G
102670-540EA 215.38 USD
102670-540 102670-542 102670-544
3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile
3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

Hazard: Toxic

Formula: C₁₀H₈N₄
Melting Pt: 155…156 °C
MDL Number: MFCD00828633
CAS Number: 138942-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR