N-(3,4-Dimethylphenyl)imidodicarbonimidic diamide

Supplier: Matrix Scientific

080637-500MG 080637-1G
102670-506EA 258.46 USD
102670-506 102670-508
N-(3,4-Dimethylphenyl)imidodicarbonimidic diamide
N-(3,4-Dimethylphenyl)imidodicarbonimidic diamide

Hazard: Irritant

Formula: C₁₀H₁₅N₅
MDL Number: MFCD03615405
CAS Number: 49872-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR