1,4-Diphenyl-1H-pyrazol-5-amine

Supplier: Matrix Scientific

080632-500MG 080632-1G
102670-480EA 258.46 USD
102670-480 102670-482
1,4-Diphenyl-1H-pyrazol-5-amine
1,4-Diphenyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₅H₁₃N₃
MDL Number: MFCD00140147
CAS Number: 108719-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR