3-Phenyl-4,5,6,7-tetrahydro-1H-indazole

Supplier: Matrix Scientific

080627-500MG 080627-1G
102670-460EA 387.69 USD
102670-460 102670-462
3-Phenyl-4,5,6,7-tetrahydro-1H-indazole
3-Phenyl-4,5,6,7-tetrahydro-1H-indazole

Hazard: Irritant

Formula: C₁₃H₁₄N₂
MDL Number: MFCD22374930
CAS Number: 28748-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR