2-Amino-4,6-dimethylpyrimidine-5-carbonitrile

Supplier: Matrix Scientific

080626-500MG 080626-1G
102670-456EA 387.69 USD
102670-456 102670-458
2-Amino-4,6-dimethylpyrimidine-5-carbonitrile
2-Amino-4,6-dimethylpyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₇H₈N₄
MDL Number: MFCD18821968
CAS Number: 16341-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR