(3E)-1-Benzyl-3-[(dimethylamino)methylene]piperidin-4-one
Supplier: Matrix Scientific
080613-500MG
080613-1G
102670-398EA
387.69
USD
102670-398
102670-400
(3E)-1-Benzyl-3-[(dimethylamino)methylene]piperidin-4-one
(3E)-1-Benzyl-3-[(dimethylamino)methylene]piperidin-4-one
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O MW: 244.34 g/mol |
MDL Number:
MFCD17015687 CAS Number: 35024-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...