6-Amino-1-(2-morpholin-4-ylethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080603-500MG 080603-1G 080603-5G
102670-346EA 258.46 USD
102670-346 102670-348 102670-350
6-Amino-1-(2-morpholin-4-ylethyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(2-morpholin-4-ylethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₀H₁₆N₄O₃
MW: 240.26 g/mol
MDL Number: MFCD09439083
CAS Number: 924834-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR