6-Amino-1-(2-methoxyethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080601-500MG 080601-1G 080601-5G
102670-336EA 387.69 USD
102670-336 102670-338 102670-340
6-Amino-1-(2-methoxyethyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(2-methoxyethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₇H₁₁N₃O₃
MDL Number: MFCD03954446
CAS Number: 56075-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR