6-Amino-1-(2-phenylethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080597-500MG 080597-1G
102670-318EA 193.85 USD
102670-318 102670-320
6-Amino-1-(2-phenylethyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(2-phenylethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₂
MDL Number: MFCD06483946
CAS Number: 54052-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR