6-Amino-1-ethylpyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080595-1G 080595-5G
102670-310EA 607.69 USD
102670-310 102670-312
6-Amino-1-ethylpyrimidine-2,4(1H,3H)-dione
6-Amino-1-ethylpyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₆H₉N₃O₂
MDL Number: MFCD00091925
CAS Number: 41862-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR