3-Mercapto-6,6-dimethyl-1,6-dihydro-1,2,4-triazin-5(4H)-one

Supplier: Matrix Scientific

080577-500MG 080577-1G
102670-248EA 387.69 USD
102670-248 102670-250
3-Mercapto-6,6-dimethyl-1,6-dihydro-1,2,4-triazin-5(4H)-one
3-Mercapto-6,6-dimethyl-1,6-dihydro-1,2,4-triazin-5(4H)-one

Hazard: Irritant

Formula: C₅H₉N₃OS
MW: 159.21 g/mol
MDL Number: MFCD00522533
CAS Number: 16992-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR