2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine

Supplier: Matrix Scientific

080572-500MG 080572-1G 080572-5G
102670-222EA 387.69 USD
102670-222 102670-224 102670-226
2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine
2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MDL Number: MFCD00183927
CAS Number: 1026-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR