N-(4,8-Dimethylquinazolin-2-yl)guanidine

Supplier: Matrix Scientific

080560-500MG 080560-1G
102670-158EA 258.46 USD
102670-158 102670-160
N-(4,8-Dimethylquinazolin-2-yl)guanidine
N-(4,8-Dimethylquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₁H₁₃N₅
MDL Number: MFCD00980620
CAS Number: 5361-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR