Ethyl-6-methyl-3-oxoheptanoate
Supplier: Matrix Scientific
080555-500MG
080555-1G
080555-5G
102670-136EA
516.92
USD
102670-136
102670-138
102670-140
Ethyl-6-methyl-3-oxoheptanoate
Ethyl-6-methyl-3-oxoheptanoate
Hazard: Irritant
Formula:
C₁₀H₁₈O₃ MW: 186.25 g/mol |
MDL Number:
MFCD03844432 CAS Number: 57689-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...