Ethyl-2-acetyl-5-methylhexanoate

Supplier: Matrix Scientific

080553-500MG 080553-1G
102670-128EA 387.69 USD
102670-128 102670-130
Ethyl-2-acetyl-5-methylhexanoate
Ethyl-2-acetyl-5-methylhexanoate

Hazard: Irritant

Formula: C₁₁H₂₀O₃
MDL Number: MFCD20133661
CAS Number: 1522-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR