Ethyl-3-oxo-2-(2-phenylethyl)butanoate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-Acetyl-4-phenylbutanoate

080552-500MG 080552-1G 080552-5G
102670-122EA 184.62 USD
102670-122 102670-124 102670-126
Ethyl-3-oxo-2-(2-phenylethyl)butanoate
Ethyl-3-oxo-2-(2-phenylethyl)butanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MW: 234.3 g/mol
MDL Number: MFCD22374908
CAS Number: 5337-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR