Ethyl-2-(3-methylbenzyl)-3-oxobutanoate

Supplier: Matrix Scientific

080541-500MG 080541-1G 080541-5G
102670-058EA 167.69 USD
102670-058 102670-060 102670-062
Ethyl-2-(3-methylbenzyl)-3-oxobutanoate
Ethyl-2-(3-methylbenzyl)-3-oxobutanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD16317680
CAS Number: 61713-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR