Ethyl-2-benzoyl-4-cyanobutanoate

Supplier: Matrix Scientific

080538-500MG 080538-1G 080538-5G
102670-042EA 258.46 USD
102670-042 102670-044 102670-046
Ethyl-2-benzoyl-4-cyanobutanoate
Ethyl-2-benzoyl-4-cyanobutanoate

Hazard: Irritant

Formula: C₁₄H₁₅NO₃
MDL Number: MFCD22374905
CAS Number: 5415-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR