Methyl-3-biphenyl-4-yl-3-oxopropanoate

Supplier: Matrix Scientific

080531-500MG 080531-1G 080531-5G
102670-002EA 193.85 USD
102670-002 102670-004 102670-006
Methyl-3-biphenyl-4-yl-3-oxopropanoate
Methyl-3-biphenyl-4-yl-3-oxopropanoate

Hazard: Irritant

Formula: C₁₆H₁₄O₃
MDL Number: MFCD03424765
CAS Number: 56216-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR