2-Amino-5-butyl-6-methylpyrimidin-4(3H)-one

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-butyl-6-methylpyrimidin-4-ol

080510-500MG 080510-1G
102669-928EA 193.85 USD
102669-928 102669-930
2-Amino-5-butyl-6-methylpyrimidin-4(3H)-one
2-Amino-5-butyl-6-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₉H₁₅N₃O
MW: 181.24 g/mol
MDL Number: MFCD00767145
CAS Number: 4038-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR