1,6-Diphenylhexane-1,3,4,6-tetrone

Supplier: Matrix Scientific

080433-500MG 080433-1G 080433-5G
102669-638EA 193.85 USD
102669-638 102669-640 102669-642
1,6-Diphenylhexane-1,3,4,6-tetrone
1,6-Diphenylhexane-1,3,4,6-tetrone

Hazard: Irritant

Formula: C₁₈H₁₄O₄
MDL Number: MFCD00187207
CAS Number: 53454-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR