1,5-Diphenyl-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1,5-diphenylpyrazole-3-carboxylic acid

080432-500MG 080432-1G 080432-5G
102669-632EA 418.46 USD
102669-632 102669-634 102669-636
1,5-Diphenyl-1H-pyrazole-3-carboxylic acid
1,5-Diphenyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₂
MW: 264.29 g/mol
MDL Number: MFCD01427273
CAS Number: 13599-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR