1,4-Dioxan-2-ylmethanol

Supplier: Matrix Scientific

Synonyms: (1,4-Dioxan-2-yl)methanol

080429-500MG 080429-1G
102669-616EA 646.15 USD
102669-616 102669-618
1,4-Dioxan-2-ylmethanol
1,4-Dioxan-2-ylmethanol

Hazard: Irritant

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD08669844
CAS Number: 29908-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR