4,5-Dimethylpyrimidin-2-amine

Supplier: Matrix Scientific

080427-500MG 080427-1G 080427-5G
102669-604EA 355.38 USD
102669-604 102669-606 102669-608
4,5-Dimethylpyrimidin-2-amine
4,5-Dimethylpyrimidin-2-amine

Hazard: Irritant

Formula: C₆H₉N₃
MDL Number: MFCD00151982
CAS Number: 1193-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR