1-Phenyl-1H-pyrrole-2-carboxylic acid

Supplier: Matrix Scientific

080426-500MG 080426-1G
102669-600EA 444.62 USD
102669-600 102669-602
1-Phenyl-1H-pyrrole-2-carboxylic acid
1-Phenyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
MDL Number: MFCD05174745
CAS Number: 78540-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR