(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide

Supplier: Matrix Scientific

Synonyms: N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide, 4-Formylaminoantipyrine

080425-500MG 080425-1G 080425-5G
102669-594EA 258.46 USD
102669-594 102669-596 102669-598
(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide
(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₂
MW: 231.26 g/mol
MDL Number: MFCD00085457
CAS Number: 1672-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR