(1-Methyl-1H-pyrazol-5-yl)acetonitrile

Supplier: Matrix Scientific

080391-500MG 080391-1G 080391-5G
102669-424EA 646.15 USD
102669-424 102669-426 102669-428
(1-Methyl-1H-pyrazol-5-yl)acetonitrile
(1-Methyl-1H-pyrazol-5-yl)acetonitrile

Hazard: Irritant

Formula: C₆H₇N₃
MDL Number: MFCD09859321
CAS Number: 1071814-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR