1-Butyl-1H-pyrazol-3-amine

Supplier: Matrix Scientific

080381-500MG 080381-1G 080381-5G
102669-368EA 516.92 USD
102669-368 102669-370 102669-372
1-Butyl-1H-pyrazol-3-amine
1-Butyl-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₇H₁₃N₃
MDL Number: MFCD06805166
CAS Number: 73616-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR