[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]amine
Supplier: Matrix Scientific
080378-500MG
080378-1G
102669-354EA
516.92
USD
102669-354
102669-356
[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]amine
[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]amine
Hazard: Irritant
Formula:
C₅H₈N₄O₂ |
MDL Number:
MFCD04967286 CAS Number: 1002033-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...