5-Amino-3-(4-fluorophenyl)isoxazole

Supplier: Matrix Scientific

080376-500MG 080376-1G 080376-5G
102669-344EA 387.69 USD
102669-344 102669-346 102669-348
5-Amino-3-(4-fluorophenyl)isoxazole
5-Amino-3-(4-fluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₇FN₂O
MDL Number: MFCD03407398
CAS Number: 81465-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR