3-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Supplier: Matrix Scientific

080353-500MG 080353-1G
102669-244EA 646.15 USD
102669-244 102669-246
3-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride
3-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₀Cl₂N₂
MDL Number: MFCD28024761
CAS Number: 1172973-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR