[1-(1-Ethyl-1H-pyrazol-3-yl)ethyl]amine

Supplier: Matrix Scientific

080338-500MG 080338-1G
102669-180EA 646.15 USD
102669-180 102669-182
[1-(1-Ethyl-1H-pyrazol-3-yl)ethyl]amine
[1-(1-Ethyl-1H-pyrazol-3-yl)ethyl]amine

Hazard: Irritant

Formula: C₇H₁₃N₃
MDL Number: MFCD04969728
CAS Number: 911788-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR