[1-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
Supplier: Matrix Scientific
080334-500MG
080334-1G
080334-5G
102669-160EA
646.15
USD
102669-160
102669-162
102669-164
[1-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
[1-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
Hazard: Irritant
Formula:
C₆H₁₁N₃ |
MDL Number:
MFCD04969725 CAS Number: 911788-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...