[1-Methyl-2-(4-methyl-1H-pyrazol-1-yl)ethyl]amine

Supplier: Matrix Scientific

080326-500MG 080326-1G 080326-5G
102669-112EA 327.69 USD
102669-112 102669-114 102669-116
[1-Methyl-2-(4-methyl-1H-pyrazol-1-yl)ethyl]amine
[1-Methyl-2-(4-methyl-1H-pyrazol-1-yl)ethyl]amine

Hazard: Irritant

Formula: C₇H₁₃N₃
MDL Number: MFCD04970165
CAS Number: 1006483-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR