[(1-Propyl-1H-pyrazol-3-yl)methyl]amine
Supplier: Matrix Scientific
080323-500MG
080323-1G
080323-5G
102669-094EA
646.15
USD
102669-094
102669-096
102669-098
[(1-Propyl-1H-pyrazol-3-yl)methyl]amine
[(1-Propyl-1H-pyrazol-3-yl)methyl]amine
Hazard: Irritant
Formula:
C₇H₁₃N₃ |
MDL Number:
MFCD04970275 CAS Number: 1006333-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...