1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridin-5-amine

Supplier: Matrix Scientific

080321-500MG 080321-1G 080321-5G
102669-082EA 646.15 USD
102669-082 102669-084 102669-086
1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridin-5-amine
1,3-Dimethyl-1H-pyrazolo[3,4-b]pyridin-5-amine

Hazard: Irritant

Formula: C₈H₁₀N₄
MDL Number: MFCD09040556
CAS Number: 19736-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR